γ-(N-Morpholino)propanesulphonic acid (MOPS), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
69947-25G 69947-500G
SIAL69947-25GEA 127 EUR
SIAL69947-25G SIAL69947-500G
γ-(N-Morpholino)propanesulphonic acid (MOPS), Sigma-Aldrich®
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
Melting Pt: > 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288