MES sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Sodium 2-Morpholinoethanesulphonate, Sodium 3-morpholin-1-ylethylsulphonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, MES-Na
M-Clarity™ quality level = MQ400
M5057-100G
M5057-25G
SIALM5057-100GEA
241
EUR
SIALM5057-100G
SIALM5057-25G
MES sodium salt, Sigma-Aldrich®
MES sodium salt
Formula:
C₆H₁₂NNaO₄S MW: 217.22 g/mol |
MDL Number:
MFCD00065473 CAS Number: 71119-23-8 EINECS: 275-203-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...