Penicillin G sodium salt, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
PENNA-100MU PENNA-10MU
SIALPENNA-100MUEA 163 EUR
SIALPENNA-100MU SIALPENNA-10MU
Penicillin G sodium salt, Sigma-Aldrich®
Penicillin G sodium salt
Formula: C₁₆H₁₇N₂NaO₄S
Melting Pt: 255 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00069666
CAS Number: 69-57-8
EINECS: 200-710-2