1-Bromo-2-iodobenzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Bromo-1-iodobenzene, 2-Bromoiodobenzene
M-Clarity™ quality level = MQ100
242616-25G 242616-5G
SIAL242616-25GEA 92.2 EUR
SIAL242616-25G SIAL242616-5G
1-Bromo-2-iodobenzene, Sigma-Aldrich®
1-Bromo-2-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 250 °C (1013 hPa)
Density: 2.2571 g/cm³ (25 °C)
MDL Number: MFCD00001030
CAS Number: 583-55-1
EINECS: 209-508-9