4-Bromo-2-nitrophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
309877-5G
SIAL309877-5GEA 31.5 EUR
SIAL309877-5G
4-Bromo-2-nitrophenol, Sigma-Aldrich®
4-Bromo-2-nitrophenol
Formula: C₆H₄BrNO₃
MW: 218.01 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 90…94 °C
MDL Number: MFCD00082540
CAS Number: 7693-52-9
EINECS: 231-707-4