2-Chloro-6-fluorophenol, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

590169-25G 590169-5G
SIAL590169-25GEA 170 EUR
SIAL590169-25G SIAL590169-5G
2-Chloro-6-fluorophenol, Sigma-Aldrich®
2-Chloro-6-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 65 °C
MDL Number: MFCD01631574
CAS Number: 2040-90-6
EINECS: 433-890-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR