Isophthaloyl dichloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A15904.22
A15904.36
A15904.22EA
36.3
EUR
A15904.22
A15904.36
Isophthaloyl dichloride ≥98%
Isophthaloyl dichloride
Purity: 98%
CAS No.: 99-63-8
Molecular Formula: C8H4Cl2O2
Formula Weight: 203.02
Flash Point: 180°(356°F)
Melting Point: 42-45°
Boiling Point: 275-278°
Beilstein Reference: 638342
UN No.: 3261
MDL No.: MFCD00000678
Formula:
C₈H₄Cl₂O₂ Boiling Pt: 275…278 °C Melting Pt: 42…45 °C Density: 1.388 Flash Pt: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000678 CAS Number: 99-63-8 EINECS: 202-774-7 UN: 3261 ADR: 8,II |