Bis(2-ethylhexyl)phthalate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Di-iso-octyl phthalate, Dioctyl phthalate, Di-(2-ethylhexyl) phthalate, DEHP, Di(2-ethylhexyl) phthalate, Phthalic acid bis(2-ethylhexyl) ester
M-Clarity™ quality level = MQ200
D201154-2.5L D201154-25L D201154-500ML D201154-5ML D201154-100ML D201154-2L
SIALD201154-2.5LEA 192 EUR
SIALD201154-2.5L SIALD201154-25L SIALD201154-500ML SIALD201154-5ML SIALD201154-100ML SIALD201154-2L
Bis(2-ethylhexyl)phthalate, Sigma-Aldrich®
Bis(2-ethylhexyl) phthalate
Formula: C₂₄H₃₈O₄
MW: 390.56 g/mol
Boiling Pt: 386.9 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00009493
CAS Number: 117-81-7
EINECS: 204-211-0
UN: 3082
ADR: 9,III
Merck Index: 13,02884
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8538f5b87da42d0456afcf02ecf7ed2073d3f6e4&vid=fe0df8cf0e9a1b142fe7092d43e1374cc2b50d6e&prid=web&clid=SAPDAM