Dimethyl isophthalate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Isophthalic acid dimethyl ester
M-Clarity™ quality level = MQ200
194239-25G
194239-3KG
194239-500G
SIAL194239-25GEA
43.5
EUR
SIAL194239-25G
SIAL194239-3KG
SIAL194239-500G
Dimethyl isophthalate, Sigma-Aldrich®
Dimethyl isophthalate
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Boiling Pt: 285 °C (1013 hPa) Melting Pt: 61…63 °C Density: 1.186 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008433 CAS Number: 1459-93-4 EINECS: 215-951-9 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...