5-Hydroxysophthalic acid 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 5-Hydroxyisophthalic acid
271390250 271391000 271395000
ACRO271390250EA 28 EUR
ACRO271390250 ACRO271391000 ACRO271395000
5-Hydroxysophthalic acid 99%
5-Hydroxysophthalic acid
Formula: C₈H₆O₅
MW: 182.13 g/mol
Boiling Pt: 386
Melting Pt: 297…303 °C
MDL Number: MFCD00002515
CAS Number: 618-83-7
EINECS: 210-565-7
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 297 to 303 °C
Titration with NaOH ≥98.5 %