2-Fluoro-4-iodoaniline ≥99%
Supplier: Thermo Fisher Scientific
Danger
B22450.14 B22450.06 B22450.22
B22450.14EA 90.3 EUR
B22450.14 B22450.06 B22450.22
2-Fluoro-4-iodoaniline ≥99%
2-Fluoro-4-iodoaniline
Purity: 99% CAS No.: 29632-74-4 Molecular Formula: C6H5FIN Formula Weight: 237.02 Flash Point: 110°(230°F) Melting Point: 55-57° Beilstein Reference: 2081100 MDL No.: MFCD00011738
Formula: C₆H₅FIN
MW: 237.02 g/mol
Boiling Pt: 259
Melting Pt: 55…57 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011738
CAS Number: 29632-74-4