2,4-Dinitrophenol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,3-Dinitro-4-hydroxybenzene, α-Dinitrophenol
M-Clarity™ quality level = MQ200
D198501-5G D198501-100G D198501-1KG
SIALD198501-5GEA 26.6 EUR
SIALD198501-5G SIALD198501-100G SIALD198501-1KG
2,4-Dinitrophenol, Sigma-Aldrich®
2,4-Dinitrophenol
Formula: C₆H₄N₂O₅
MW: 184.11 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 132…133 °C
Density: ∼1.675 g/cm³ (20 °C)
CAS Number: 51-28-5
EINECS: 200-087-7
UN: 1320
ADR: 4.1,I
Merck Index: 13,03309