Phenol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

M-Clarity™ quality level = MQ100

P1037-100G P1037-25G P1037-500G
SIALP1037-100GEA 68.1 EUR
SIALP1037-100G SIALP1037-25G SIALP1037-500G
Phenol, Sigma-Aldrich®
Phenol
Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Density: 1.0722 g/cm³ (20 °C)
Flash Pt: 81
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
REACH: 01-2119471329-32
Merck Index: 12,07390

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR