ω-Bromo-4-cyanoacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-(2-Bromoacetyl)benzonitrile, 4-(Bromoacetyl)benzonitrile, 4-(2-Bromoacetyl)benzenecarbonitrile, 2-Bromo-4'-cyanoacetophenone, Bromomethyl p-cyanophenyl ketone, 2-Bromo-4′-cyanoacetophenone, 4-Cyanophenacyl bromide, α-Bromo-4-cyanoacetophenone
M-Clarity™ quality level = MQ100
539392-25G
539392-5G
SIAL539392-25GEA
82.2
EUR
SIAL539392-25G
SIAL539392-5G
ω-Bromo-4-cyanoacetophenone, Sigma-Aldrich®
ω-Bromo-4-cyanoacetophenone
Formula:
C₉H₆BrNO MW: 224.06 g/mol Boiling Pt: 314 Melting Pt: 92…94 °C |
MDL Number:
MFCD00052931 CAS Number: 20099-89-2 UN: 2923 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...