ω-Bromo-4-nitroacetophenone 95%
Supplier: Thermo Scientific
Danger
Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone
305480100 305480500
ACRO305480100EA 37.8 EUR
ACRO305480100 ACRO305480500
ω-Bromo-4-nitroacetophenone 95%
ω-Bromo-4-nitroacetophenone
Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Melting Pt: 94…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007356
CAS Number: 99-81-0
EINECS: 202-789-9
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 100 °C
GC ≥94 %