ω-Bromo-3-fluoroacetophenone
Supplier: Thermo Scientific
Synonyms: Bromomethyl m-fluorophenyl ketone, 3-Fluorophenacyl bromide, 2-Bromo-1-(3-fluorophenyl)ethan-1-one, α-Bromo-3-fluoroacetophenone, 2-Bromo-3'-fluoroacetophenone
SPB06322DA SPB06322EA
MAYBSPB06322DAEA 0 EUR
MAYBSPB06322DA MAYBSPB06322EA
ω-Bromo-3-fluoroacetophenone
ω-Bromo-3-fluoroacetophenone
Formula: C₈H₆BrFO
MW: 217.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00109756
CAS Number: 53631-18-8