2',6'-Dichloroacetophenone, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-(2,6-Dichlorophenyl)ethanone

M-Clarity™ quality level = MQ200

593753-1G 593753-5G
SIAL593753-1GEA 31.5 EUR
SIAL593753-1G SIAL593753-5G
2',6'-Dichloroacetophenone, Sigma-Aldrich®
2',6'-Dichloroacetophenone
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
MDL Number: MFCD00016339
CAS Number: 2040-05-3
EINECS: 218-035-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR