2'-Hydroxy-5'-methoxyacetophenone 99
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2'-Hydroxy-5'-Methoxyacetophenone, 2′-Hydroxy-5′-methoxyacetophenone
A19216.06 A19216.14
A19216.06EA 65.7 EUR
A19216.06 A19216.14
2'-Hydroxy-5'-methoxyacetophenone 99
2'-Hydroxy-5'-methoxyacetophenone
Purity: 99% CAS No.: 705-15-7 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 48-50° Beilstein Reference: 1239364 MDL No.: MFCD00008731
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281
Melting Pt: 48…50 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008731
CAS Number: 705-15-7
EINECS: 211-882-3