2'-Hydroxy-6'-methoxyacetophenone ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-(2-Hydroxy-6-methoxyphenyl)ethanone
A18539.03 A18539.06 A18539.14
A18539.03EA 49.1 EUR
A18539.03 A18539.06 A18539.14
2'-Hydroxy-6'-methoxyacetophenone ≥97%
2'-Hydroxy-6'-methoxyacetophenone
Purity: 97% CAS No.: 703-23-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 56-59° Boiling Point: 141°/16mm MDL No.: MFCD00008732
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 141 °C (16 mmHg)
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00008732
CAS Number: 703-23-1
EINECS: 211-872-9