1-Phenyl-1,2-propanedione, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Phenylpropane-1,2-dione
M-Clarity™ quality level = MQ300
W322601-100G-K W322601-1KG-K W322601-5KG-K
SIALW322601-100G-KEA 98.9 EUR
SIALW322601-100G-K SIALW322601-1KG-K SIALW322601-5KG-K
1-Phenyl-1,2-propanedione, Sigma-Aldrich®
1-Phenyl-1,2-propanedione
Formula: C₆H₅C(O)C(O)CH₃
MW: 148.16 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: ? 20 °C
Density: 1.116 g/cm³ (20 °C)
MDL Number: MFCD00008755
CAS Number: 579-07-7
EINECS: 209-435-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1394d5392b739ebd2496078378d299949ea575ae&vid=b85d2a588e386bc41f5caca908cd1530a3da9131&prid=web&clid=SAPDAM