2-Fluoro-5-(trifluoromethyl)phenyl isocyanate 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Fluoro-5-(trifluoromethyl)phenylisocyanate

AC31934.250 AC31934.1
MAYBAC31934.250EA 0 EUR
MAYBAC31934.250 MAYBAC31934.1
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate 97%
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
Formula: C₈H₃F₄NO
MW: 205.11 g/mol
Boiling Pt: 53 °C (2.5 mmHg)
Density: 1,418 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00673069
CAS Number: 69922-27-6
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4560 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR