α,α-Dimethoxytoluene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Benzaldehyde dimethyl acetal
M-Clarity™ quality level = MQ200
226076-100G 226076-500G
SIAL226076-100GEA 50.5 EUR
SIAL226076-100G SIAL226076-500G
α,α-Dimethoxytoluene, Sigma-Aldrich®
α,α-Dimethoxytoluene
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 198
Density: 1.01 g/cm³ (20 °C)
MDL Number: MFCD00008491
CAS Number: 1125-88-8
EINECS: 214-413-0