2-tert-Butyl-p-cresol ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-tert-Butyl-2-hydroxy-5-methylbenzene, 2-tert-Butyl-4-methylphenol, 3-tert-Butyl-4-hydroxytoluene
B23545.22
B23545.36
B23545.22EA
31.5
EUR
B23545.22
B23545.36
2-tert-Butyl-p-cresol ≥99%
2-tert-Butyl-p-cresol
Purity: 99%
CAS No.: 2409-55-4
Molecular Formula: C11H16O
Formula Weight: 164.25
Flash Point: 105°(221°F)
Melting Point: 49-54°
Boiling Point: 237°
Beilstein Reference: 1817645
UN No.: 2430
MDL No.: MFCD00002381
Formula:
C₁₁H₁₆O Boiling Pt: 237 °C Melting Pt: 49…54 °C Density: 0.961 g/cm³ (20 °C) Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002381 CAS Number: 2409-55-4 EINECS: 219-314-6 UN: 2430 ADR: 8,III |