1-Iodo-2,3,4,5-tetramethylbenzene
Supplier: Thermo Scientific
CD07685.1
CD07685.10
CD07685.50
MAYBCD07685.1EA
0
EUR
MAYBCD07685.1
MAYBCD07685.10
MAYBCD07685.50
1-Iodo-2,3,4,5-tetramethylbenzene
1-Iodo-2,3,4,5-tetramethylbenzene
Formula:
C₁₀H₁₃I MW: 260.12 g/mol |
CAS Number:
54509-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...