3-Quinolinecarboxylic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

177148-1G 177148-25G 177148-5G
SIAL177148-1GEA 32.4 EUR
SIAL177148-1G SIAL177148-25G SIAL177148-5G
3-Quinolinecarboxylic acid, Sigma-Aldrich®
3-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 282…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006770
CAS Number: 6480-68-8
EINECS: 229-337-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR