1,2-Epoxycyclohexane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Epoxycyclohexane, Cyclohexene oxide
M-Clarity™ quality level = MQ100
C102504-100ML C102504-25ML C102504-500ML
SIALC102504-100MLEA 56.5 EUR
SIALC102504-100ML SIALC102504-25ML SIALC102504-500ML
1,2-Epoxycyclohexane, Sigma-Aldrich®
1,2-Epoxycyclohexane
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –27 °C
Density: 0.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005162
CAS Number: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8,II