1-Acetylcyclohexene 97%
Supplier: Thermo Fisher Scientific
Synonyms: 1-Acetyl-1-cyclohexene, Cyclohex-1-enylmethylketone, 1-(Cyclohex-1-en-1-yl)ethanone
102500050 102500250
ACRO102500050EA 36.5 EUR
ACRO102500050 ACRO102500250
1-Acetylcyclohexene 97%
1-Acetylcyclohexene
Formula: C₈H₁₂O
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 73 °C
Density: 0.96 g/cm³
Flash Pt: 65 °C
MDL Number: MFCD00001547
CAS Number: 932-66-1
EINECS: 213-256-5
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4920 (20°C, 589 nm)
GC ≥96.0 %