Diphenylcyclopropenone, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one

M-Clarity™ quality level = MQ100

177377-1G 177377-5G
SIAL177377-1GEA 72.6 EUR
SIAL177377-1G SIAL177377-5G
Diphenylcyclopropenone, Sigma-Aldrich®
Diphenylcyclopropenone
Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 407
Melting Pt: 112…113 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00001311
CAS Number: 886-38-4
EINECS: 212-948-4
Merck Index: 12,03366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR