PIPES sesquisodium salt

Supplier: Roth Carl

ROTH5583.1EA 0 EUR
ROTH5583.1 ROTH5583.2
PIPES sesquisodium salt
PIPES sesquisodium salt
Formula: C₁₆H₃₃N₄Na₃O₁₂S₄
MW: 670,69 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00066393
CAS Number: 100037-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR