1-(2-Methoxyphenyl)piperazine
Supplier: Thermo Scientific
RH01755.1 RH01755.10 RH01755.50
MAYBRH01755.1EA 0 EUR
MAYBRH01755.1 MAYBRH01755.10 MAYBRH01755.50
1-(2-Methoxyphenyl)piperazine
1-(2-Methoxyphenyl)piperazine
Formula: C₁₁H₁₆N₂O
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 35…38 °C
Density: 1.09 g/cm³ (20 °C)
MDL Number: MFCD00005958
CAS Number: 35386-24-4
EINECS: 252-537-7
UN: 2923
ADR: 8,III