Phenothiazine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 10H-Phenothiazine
M-Clarity™ quality level = MQ200
P14831-1KG P14831-25G P14831-500G
SIALP14831-1KGEA 139 EUR
SIALP14831-1KG SIALP14831-25G SIALP14831-500G
Phenothiazine, Sigma-Aldrich®
Phenothiazine
Formula: C₁₂H₉NS
MW: 199.28 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 182…185 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: 202 °C
MDL Number: MFCD00005015
CAS Number: 92-84-2
EINECS: 202-196-5
Merck Index: 12,07404