Lithium perchlorate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
634565-100G 634565-10G
SIAL634565-100GEA 368 EUR
SIAL634565-100G SIAL634565-10G
Lithium perchlorate, Sigma-Aldrich®
Lithium perchlorate
Formula: ClLiO₄
MW: 106.39 g/mol
Boiling Pt: 430
Melting Pt: 236…237 °C
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011079
CAS Number: 7791-03-9
EINECS: 232-237-2
UN: 1481
ADR: 5.1 II
Merck Index: 13,05561