Potassium molybdate(VI), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Potassium molybdate, Molybdic acid potassium salt, Dipotassium tetraoxomolybdate, Molybdic acid, potassium salt, Potassium molybdenum oxide, di-Potassium molybdate
MQ100
308390-100G 308390-500G
SIAL308390-100GEA 89.1 EUR
SIAL308390-100G SIAL308390-500G
Potassium molybdate(VI), Sigma-Aldrich®
Potassium molybdate(VI)
Formula: K₂MoO₄
MW: 238.13 g/mol
Melting Pt: 926
Density: 2.91 g/cm³ (18 °C)
MDL Number: MFCD00011370
CAS Number: 13446-49-6
EINECS: 236-599-2
Merck Index: 13,07732