Lithium hexafluorophosphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Lithium hexafluorophosphate(V)
M-Clarity™ quality level = MQ100
450227-25G 450227-5G
SIAL450227-25GEA 341 EUR
SIAL450227-25G SIAL450227-5G
Lithium hexafluorophosphate, Sigma-Aldrich®
Lithium hexafluorophosphate
Formula: F₆LiP
MW: 151.91 g/mol
Density: ∼1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011096
CAS Number: 21324-40-3
EINECS: 244-334-7
UN: 3260
ADR: 8 I