Disodium hydrogen phosphate
Supplier: Roth Carl
T876.1 T876.2 T876.3
ROTHT876.1EA 38.7 EUR
ROTHT876.1 ROTHT876.2 ROTHT876.3
Disodium hydrogen phosphate
di-Sodium hydrogen phosphate
Formula: Na₂HPO₄
Melting Pt: 250 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805