Potassium titanate
Supplier: Thermo Fisher Scientific
B20263.22 B20263.36
B20263.22EA 51.5 EUR
B20263.22 B20263.36
Potassium titanate
Potassium titanate
CAS No.: 12030-97-6 Molecular Formula: K2O + TiO2 Odour: Odourless Melting Point: 1615° MDL No.: MFCD00058803
Formula: K₂O₃Ti
Melting Pt: 1615 °C
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00058803
CAS Number: 12030-97-6
EINECS: 234-748-6