Calcium metasilicate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Calcium silicate
M-Clarity™ quality level = MQ100
372668-250G 372668-50G
SIAL372668-250GEA 474 EUR
SIAL372668-250G SIAL372668-50G
Calcium metasilicate, Sigma-Aldrich®
Calcium metasilicate
Formula: CaO₃Si
MW: 116.16 g/mol
MDL Number: MFCD00015979
CAS Number: 10101-39-0
EINECS: 233-250-6