Barium titanate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
M-Clarity™ quality level = MQ200
338842-100G 338842-500G
SIAL338842-100GEA 89.5 EUR
SIAL338842-100G SIAL338842-500G
Barium titanate, Sigma-Aldrich®
Barium titanate
Formula: BaO₃Ti
MW: 233.21 g/mol
Melting Pt: 1625
Density: 5.67 g/cm³ (20 °C)
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1 III
Merck Index: 12,01029