Boron triiodide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
307629-25G
307629-5G
SIAL307629-25GEA
1440
EUR
SIAL307629-25G
SIAL307629-5G
Boron triiodide, Sigma-Aldrich®
Boron triiodide
Formula:
BI₃ MW: 391.52 g/mol Boiling Pt: 210 °C (1013 hPa) Melting Pt: 43 °C Density: 3.35 g/cm³ (20 °C) |
MDL Number:
MFCD00036286 CAS Number: 13517-10-7 EINECS: 236-857-4 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...