Gallium nitride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Gallium nitride, Gallium(III) nitride
M-Clarity™ quality level = MQ100
481769-10G 481769-50G
SIAL481769-10GEA 265 EUR
SIAL481769-10G SIAL481769-50G
Gallium nitride, Sigma-Aldrich®
Gallium nitride
Formula: GaN
MW: 83.73 g/mol
Melting Pt: 800
Density: 6.1 g/cm³ (20 °C)
MDL Number: MFCD00016108
CAS Number: 25617-97-4
EINECS: 247-129-0