Indium (III) oxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Indium trioxide, Diindium trioxide
M-Clarity™ quality level = MQ100
632317-25G 632317-5G
SIAL632317-25GEA 365 EUR
SIAL632317-25G SIAL632317-5G
Indium (III) oxide, Sigma-Aldrich®
Indium (III) oxide
Formula: In₂O₃
MW: 277.64 g/mol
Melting Pt: 715.05 °C
Density: 7.18 g/cm³ (20 °C)
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9
Merck Index: 12,04984