Lead(II,IV) oxide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Minium, Orange lead
M-Clarity™ quality level = MQ200
241547-100G
241547-500G
SIAL241547-100GEA
28.4
EUR
SIAL241547-100G
SIAL241547-500G
Lead(II,IV) oxide, Sigma-Aldrich®
Lead (II,IV) oxide
Formula:
Pb₃O₄ MW: 685.6 g/mol Boiling Pt: 2682 Melting Pt: 830 Density: 9.1 g/cm³ (20 °C) |
MDL Number:
MFCD00078491 CAS Number: 1314-41-6 EINECS: 215-235-6 UN: 1479 ADR: 5.1 II Merck Index: 12,05449 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...