Lead(II) acetate basic
Supplier: Thermo Scientific
Danger
Synonyms: Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
A16763.0E A16763.36
A16763.0EEA 317 EUR
A16763.0E A16763.36
Lead(II) acetate basic
Lead (II) acetate basic
CAS No.: 1335-32-6 Molecular Formula: C4H6O4Pb.2Pb(OH)2 Formula Weight: 807.72 Merck Reference: 14,5419 UN No.: 1616 MDL No.: MFCD00013041
Formula: (CH₃COO)₂Pb·2Pb(OH)₂
MW: 807.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011154
CAS Number: 1335-32-6
EINECS: 215-630-3
UN: 1616
ADR: 6.1,III
Merck Index: 14,5419