Antimony(III) oxide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Antimony trioxide, Diantimony trioxide, di-Antimony trioxide
M-Clarity™ quality level = MQ100
637173-100G
637173-25G
SIAL637173-100GEA
148
EUR
SIAL637173-100G
SIAL637173-25G
Antimony(III) oxide, Sigma-Aldrich®
Antimony (III) oxide
Formula:
O₃Sb₂ MW: 291.52 g/mol Boiling Pt: 1425 Melting Pt: 654 Density: 5.2 g/cm³ (20 °C) |
MDL Number:
MFCD00011214 CAS Number: 1309-64-4 EINECS: 215-175-0 UN: 1549 ADR: 6.1 III Merck Index: 13,00717 |