1,2-Bis(diphenylphosphino)ethane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)
M-Clarity™ quality level = MQ200
106496-10G 106496-50G
SIAL106496-10GEA 58.4 EUR
SIAL106496-10G SIAL106496-50G
1,2-Bis(diphenylphosphino)ethane, Sigma-Aldrich®
1,2-Bis(diphenylphosphino)ethane
Formula: C₂₆H₂₄P₂
MW: 398.42 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00003047
CAS Number: 1663-45-2
EINECS: 216-769-2