1,3-Bis(diphenylphosphino)propane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane
M-Clarity™ quality level = MQ200
262048-1G 262048-25G 262048-5G
SIAL262048-1GEA 31.1 EUR
SIAL262048-1G SIAL262048-25G SIAL262048-5G
1,3-Bis(diphenylphosphino)propane, Sigma-Aldrich®
1,3-Bis(diphenylphosphino)propane
Formula: C₂₇H₂₆P₂
MW: 412.45 g/mol
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00003050
CAS Number: 6737-42-4
EINECS: 229-791-2