Ammonium metavanadate ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Ammonium monovanadate, Ammonium vanadium trioxide, Ammonium vanadium oxide
11091.30 11091.A1
11091.30EA 127 EUR
11091.30 11091.A1
Ammonium metavanadate ≥99% (metals basis)
Ammonium metavanadate
Purity: 99% CAS No.: 7803-55-6 Molecular Formula: NH4VO3 Formula Weight: 116.98 Physical Form: Powder Density: 2.33 Melting Point: 200° dec. Merck Reference: 14,568 UN No.: 2859 MDL No.: MFCD00011430
Formula: NH₄VO₃
MW: 116.98 g/mol
Melting Pt: 200 °C (dec.)
Density: 2.33
Storage Temperature: Ambient
MDL Number: MFCD00011430
CAS Number: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1 II
Merck Index: 14,568