Tellurium dioxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
204552-10G 204552-50G
SIAL204552-10GEA 93.4 EUR
SIAL204552-10G SIAL204552-50G
Tellurium dioxide, Sigma-Aldrich®
Tellurium dioxide
Formula: O₂Te
MW: 159.6 g/mol
Boiling Pt: 1245
Melting Pt: 773
Density: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,09123