3-Hydroxyindole
Supplier: Thermo Scientific
AC35046.250
AC35046.1
MAYBAC35046.250EA
0
EUR
MAYBAC35046.250
MAYBAC35046.1
3-Hydroxyindole
3-Hydroxyindole
Formula:
C₈H₇NO MW: 133.15 g/mol |
CAS Number:
480-93-3 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...