1-Hydroxy-2-naphthoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Hydroxy-naphthalene-2-carboxylic acid
MQ200
109630-100G 109630-5G
SIAL109630-100GEA 58.5 EUR
SIAL109630-100G SIAL109630-5G
1-Hydroxy-2-naphthoic acid, Sigma-Aldrich®
1-Hydroxy-2-naphthoic acid
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Boiling Pt: 375
Melting Pt: 205
MDL Number: MFCD00003960
CAS Number: 86-48-6
EINECS: 201-674-0
UN: 2811
ADR: 6.1 III
Merck Index: 13,04859